Documenti fondamentali
BR82221
BRAND® needle-valve stopcocks for burettes and automatic burettes with PTFE spindle
Straight, for burette capacity 25 mL
Sinonimo/i:
Accessories, Burettes, Needle-valve stopcock, Stopcocks
About This Item
Codice UNSPSC:
40141602
NACRES:
NB.43
Materiali
polystyrene
Caratteristiche
for burette capacity 25 mL
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 82221
Descrizione generale
Interchangeable PTFE valve spindle, PP cap and retention device. Valve opening 0-25.5 mm.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)