BR781510
Brand®1.2 ML Tubes
with 12 strips of 8 tubes, polypropylene
Sinonimo/i:
Sample Storage, Tube racks, Tubes
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.22
Prodotti consigliati
Materiali
polypropylene
Sterilità
non-sterile
Caratteristiche
with 12 strips of 8 tubes
Produttore/marchio commerciale
BRAND 781510
Confezionamento
pkg of (10 racks with 12 strips of 8 tubes)
Capacità
1.2 mL
Categorie correlate
Descrizione generale
PP. Tubes and rack are autoclavable at 121 °C (2 bar), acc. DIN EN 285. Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA. Designed for storing and freezing down to -80 °C, and for transporting reagents and samples. Compatible with standard microplates and suitable for working with multichannel pipettes. (Size of tubes: outer Ø 8.8 mm, height 45 mm.) Single caps and strip caps, PE, available as accessories.BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Caratteristiche e vantaggi
- BRAND® 1.2 ML tubes racks are manufactured by high-quality polypropylene.
- Designed for storing and transporting reagents.
- Suitable for biological tests, such as PCR, cell growth studies, RIA, and EIA.
- Tubes and racks are autoclavable at 121°C.
- Temperature resistant down to -80°C.
- Tight sealing closure options prevents sample loss.
- Compact design is ideal for use in automated or multi-channel systems.
Note legali
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR781510-1PK | 4061831860283 |