BR701964
BRAND® repair set for TRANSFERPETTOR
for 1/2/3/4/5 μL
Sinonimo/i:
Accessories, Pipettes, Piston-operated pipettes, Positive displacement principle, Repair set, Transferpettor
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.01
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 701964
Categorie correlate
Descrizione generale
Includes: 1 allen key, 1 piston rod with fitted PTFE Transferpettor-seal (above size 20 μL), 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 Transferpettor-seals, PTFE 1 mounting block (above size 20 μL).
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)