Documenti fondamentali
BR4630170
BRAND® Dispensette® S Organic, Analog-adjustable bottle-top dispenser
volume range 10-100 mL, without recirculation valve
Sinonimo/i:
BRAND Dispensette S
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Caratteristiche
incremental 1 mL
DE-M marking
without recirculation valve
Produttore/marchio commerciale
BRAND 4630170
Intervallo di volume
10-100 mL
Raccordi
(GL 32-33)
GL 38
S 40
Descrizione generale
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Caratteristiche e vantaggi
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Altre note
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.