Documenti fondamentali
BR4600131
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.5-5 mL, with recirculation valve
Sinonimo/i:
bottle top dispensers
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Caratteristiche
incremental 0.1 mL
DE-M marking
with recirculation valve
Produttore/marchio commerciale
BRAND 4600131
Intervallo di volume
0.5-5 mL
Raccordi
(GL 28/S28)
GL 32
GL 38
S 40
Descrizione generale
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Caratteristiche e vantaggi
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Altre note
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.