Documenti fondamentali
BR30900
BRAND® rotary pipette stand for 94 pipettes
D × H 230 mm × 450 mm
Sinonimo/i:
Pipette Trays, Trays
About This Item
Codice UNSPSC:
41122801
NACRES:
NB.51
Materiali
polypropylene
Caratteristiche
Holds 94 x pipettes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 30900
D × alt.
230 mm × 450 mm
Descrizione generale
The BRAND® Rotary Pipette Stand is designed to accommodate up to 94 pipettes of various sizes, providing an organized and efficient solution for laboratory environments. Constructed from durable polypropylene (PP), this rotary model features an additional rotary tip reset mechanism that protects pipette tips from damage and contamination. It is suitable for all graduated and one-mark pipettes, ensuring versatility in use.The stand′s slightly curved base rim helps retain dripping liquids, keeping your workspace clean and organized. This practical pipette stand enhances accessibility and efficiency, making it an essential accessory for researchers and technicians in any lab setting.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)