BR23866
BRAND® spare burette length for compact automatic burette
25 mL, SILBERBRAND
Sinonimo/i:
Accessories, Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
About This Item
Codice UNSPSC:
41121808
NACRES:
NB.43
Prodotti consigliati
Materiali
amber AR-Glass
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 23866
Campo di misura
25 mL
Lungh. buretta
400 mm
Volume
25 mL
Categorie correlate
Descrizione generale
The BRAND® spare burette length for the compact automatic burette is made from high-quality Boro 3.3 clear glass, ensuring durability and clarity for precise measurements. Designed specifically for compatibility with BLAUBRAND® and SILBERBRAND compact automatic burettes, this spare part maintains the performance standards required for accurate liquid dispensing.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR23866-1EA | 4061831854961 |