BR130320
BRAND® screw cap
thread for GL W 32
Sinonimo/i:
Accessories, Bottles, Screw caps
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Materiali
polypropylene
Descrizione
for wide neck bottles
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 130320
Raccordi
thread for GL W 32
Categorie correlate
Descrizione generale
The BRAND® Screw Cap is a secure blue cap designed for square bottles, featuring a tamper-proof closure that ensures the integrity of the contents. This cap includes a press-off retaining ring and a sealing cone, providing an effective seal to prevent leaks and contamination. Its durable construction makes it suitable for various laboratory and industrial applications, ensuring reliable performance in sample storage and handling.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR130320-1EA | 4061831852783 |