Documenti fondamentali
BAH369400010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 1.25 mL , ¼tsp, L × W 7 in. (17.8 cm) × 0.8125 in. (20 mm), sterile, pack of 10 ea (individually wrapped)
Sinonimo/i:
Lab Scoop, dispensing tool, spoon
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
Materiali
white polystyrene
Sterilità
sterile
Confezionamento
pack of 10 ea (individually wrapped)
Produttore/marchio commerciale
Bel-Art H36940-0010
Lungh. × largh.
7 in. (17.8 cm) × 0.8125 in. (20 mm)
Capacità nominale
1.25 mL , ¼tsp
Descrizione generale
Designed to gather small amounts of sample material, especially in hard to reach containers.
Caratteristiche e vantaggi
  • Long handle reaches into jars, bottles and other confined areas
  • Minimizes the potential of personal contact with sampled material
  • Sterile, FDA grade polystyrene safe for food, drug and cosmetic use
  • Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)