Merck
GC Analysis of Pyrazines in Peanut Butter on SUPELCOWAX 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber
GC Analysis of Pyrazines in Peanut Butter on SUPELCOWAX 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber suitable for GC, application for SPME
材料
分析柱
产品编号
说明
价格
SUPELCOWAX 10毛细管GC色谱柱
L × I.D. 30 m × 0.25 mm, df 0.25 μm
固相微萃取纖維
产品编号
说明
价格
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS)
needle size 24 ga, for use with manual holder
標準
产品编号
说明
价格
二硫化碳
anhydrous, ≥99%
2-乙基吡嗪
≥98%, FG
2-甲基吡嗪
Pharmaceutical Secondary Standard; Certified Reference Material
二甲基二硫化物
analytical standard
2,3-二乙基吡嗪
analytical reference material
2,5-二甲基吡嗪
analytical standard
相關產品
产品编号
说明
价格
SPME 光纤支架
for use with manual sampling
SPME 光纤支架
for use with CTC CombiPAL and Gerstel MPS2
CONDITIONS
sample/matrix
5 g peanut butter in 40 mL vial
SPME fiber
divinylbenzene-Carboxen-polydimethylbenzene StableFlex (57328-U)
extraction
headspace, 30 min at 65 °C in heating block
desorption process
270 °C for 5 min
column
SUPELCOWAX 10, 30 m × 0.25 mm, 0.25 μm (24079)
oven
40 °C (5 min) to 230 °C at 4 °C/min
inj. temp.
270 °C
injection
splitless/split, closed 0.5 min, 270 °C, with 0.75 mm liner
detector
ion trap mass spectrometer, m/z =30-350 at 0.6 sec/scan. Selected ions used for quantitation.
carrier gas
Helium, 30 cm/sec
liner
0.75 mm
sample
peanut butter
说明
分析報告
In this application, SPME was used for the analysis of volatiles in multifloral honey using a headspace method. Headspace SPME (HS-SPME) using a Carboxen/PDMS/DVB fiber coupled with gas chromatography-mass spectrometry (GC/MS) was used for the characterization of the volatile fraction of honey samples. Using SPME it was possible to obtain and distinguish the chemical fingerprints of different types of honey. The relatively polar SUPELCOWAX 10 GC column provides excellent separation of these polar compounds.