技術
HPLC: suitable
測試參數
column: CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase: (A) 20 mM ammonium acetate, pH 4.1; (B) methanol; (70:30, A:B)
flow rate: 0.9 mL/min
column temp.: 25 °C
detector: UV, 254 nm
injection: 0.5 μL
sample: 2 mg/mL in methanol
適合性
application for HPLC
應用
life science and biopharma
法律資訊
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC