Merck
27473
Supelco
带 PTFE/橡胶隔垫的 11mm 钳口密封盖
red aluminum seal, seal diam. 11 mm, PTFE/red rubber septum, pkg of 1000 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121821
材料
PTFE/red rubber septum
red aluminum seal
特點
closure type crimp seal
包裝
pkg of 1000 ea
密封圈直徑
11 mm
接頭
thread for 11 mm
正在寻找类似产品? 访问 产品对比指南
Choose from one of the most recent versions:
分析证书(COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客户支持