主要文件
Z743507
Direct Thermal WetGrip labels
roll of 1000
登录查看公司和协议定价
About This Item
分類程式碼代碼:
55121611
NACRES:
NB.35
包裝
roll of 1000
製造商/商標名
Diversified Biotech GRDT-3000
長度 × 寬度
1.50 in. × 0.75 in.
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
GRIPS - To wet, frost-covered and frozen surfaces; temperature resistant to -80°C
THIN - Compatible with centrifuge rotors
RESISTANT - To chemicals and solvents
MULTI - SURFACE - Adheres to plastics, glass, metals and even paperboard boxes
For use with the GoDEX DT200 Direct printer and Dymo LabelWriter 450 (DYMO-550 printer is NOT compatible)
免責聲明
Important note for International Customers: Our Direct Thermal labels are designed to work with U.S. Model Dymo 450 printers only. They are not compatible with European Dymo printers or Dymo 550 printers.
历史批次信息供参考:
分析证书(COA)