主要文件
Z741674
Sartorius VALUEPAK pipette tips
volume range -1,200 μL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121600
最大體積:
1.2 mL
容積範圍:
-1,200 μL
無菌:
non-sterile
特點:
DNase free
RNase free
barrier: no
endotoxin free
包裝:
pack of 960 ea (refil)
pkg of refill
製造商/商標名:
Sartorius 791202
材料
colorless
無菌
non-sterile
特點
DNase free
RNase free
barrier: no
endotoxin free
包裝
pack of 960 ea (refil)
pkg of refill
製造商/商標名
Sartorius 791202
最大體積
1.2 mL
容積範圍
-1,200 μL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Large value packs of tips with the perfect tip fitting and sealing to the pipetter. This gives you the best possible accuracy and precision at the best possible price. Compatible with Optiload feature in Biohit Picus, eLINE®, mLINE® and Proline® Plus pipetters, enabling ergonomic and light tip attachment and ejection.
其他說明
Browse all pipette tips
法律資訊
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
历史批次信息供参考:
分析证书(COA)