Z663735
1 of 10
Millipore
1.10274
Sterikon® Plus 生物指示剂
Sigma-Aldrich
74041
无菌指示剂(蒸汽灭菌)
BR61750
BRAND®灭菌指示胶带
Z553964
精密密封®橡胶胶垫
1.11499
Geobacillus stearothermophilus Spore Suspension
Z553956
T8787
Triton™ X-100
1.10649
Bacillus subtilis (BGA) spore suspension
Supelco
57030-U
Visiprep™ SPE固相萃取真空装置,不含PFAS
Z553980
vinyl white
roll of 100 labels
Brady PTL-75-427
available only in EU
66.68 mm × 25.40 mm
正在寻找类似产品? 访问 产品对比指南
实验室标签