主要文件
Z374415
Sigma-Aldrich
Scienceware® microscope slide dispenser
Holds approximately 72 slides
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41100000
NACRES:
MA.01
包裝
pack of 1 ea
製造商/商標名
Bel-Art F44562-0000
ABS塑膠點膠機直徑× 寬度× 高度
4 3/8 in. (111 mm) × 4 7/8 in. (124 mm) × 5 in. (140 mm)
一般說明
Clean and dust-free. Turning the knob either direction dispenses one slide at a time.
法律資訊
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)