主要文件
Z362794
Sigma-Aldrich
Nalgene® Polymethylpentene Erlenmeyer flask
capacity 125 mL
别名:
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
colorless polypropylene cap
flat bottom
polymethylpentene
wide-neck
接合處
Joints neck (33)
無菌
non-sterile
特點
closure type Screw cap
with cap
包裝
pack of 6
製造商/商標名
Nalgene 4109-0125
容量
125 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Excellent combination of autoclavability, chemical resistance, impact strength, and clarity. Transparent to microwave radiation. Polypropylene screw closure.
其他說明
法律資訊
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
历史批次信息供参考:
分析证书(COA)