主要文件
G3900
Sigma-Aldrich
Gaskets for vertical standard gel
16.5 cm wide, size 1.5 mm × 14.5 cm, color-coded blue
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41105305
NACRES:
NB.51
产品名称
Gaskets for vertical standard gels, 16.5 cm wide, size 1.5 mm × 14.5 cm, color-coded blue
尺寸
1.5 mm × 14.5 cm
品質等級
顏色
color-coded blue
正在寻找类似产品? 访问 产品对比指南
特點和優勢
Unique silicone rubber gasket provides continuous seal around sides and bottom of glass plates, eliminating the need for a bottom spacer. Order gasket thickness to match comb and spacers. Gaskets are color-coded by thickness for easy identification. Length given is height of assembly; actual gasket length is 2 × h + w.
从最新的版本中选择一种:
分析证书(COA)