特點
holds 44 tubes (12-15 mm diam.)
製造商/商標名
Eppendorf® M1289-0020 (Global)
尺寸
360x80x129 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc.
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Global Trade Item Number
货号 | GTIN |
---|---|
EPM1289-0020-1EA | 4061835081332 |