Merck
EP6731010055
Eppendorf® CellXpert® C170i CO2 Incubator
new
inner door with 4 door segments, handle left side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41104405
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 6731010055
正在寻找类似产品? 访问 产品对比指南
一般說明
CellXpert® C170i, inner door with 4 door segments, handle left side, 100 – 127 V/50 – 60 Hz (US/JP/South America)
特點和優勢
  • handle left side
  • inner door with 4 door segments
  • 180° HTD (standard with C170i)
  • including water tray and four shelves (stainless steel)
  • 100 – 127 V, 50 – 60 Hz
法律資訊
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
从最新的版本中选择一种:
分析证书(COA)