BAH378620202
1 of 10
BAF441710000
Microscope View-Pack®
Millipore
SLSV025NB
Millex® PVDF 针式过滤器
SLCR033
Millex®亲水PTFE针式过滤器
BAF134730000
Bel-Art® Write-on™ Tape Dispenser
Sigma-Aldrich
126925
酵母菌乙醛脱氢酶
Roche
10171832001
醛脱氢酶
71077
X-Gal溶液 - Novagen
Supelco
1.04015
玻璃珠3 mm
A6338
乙醛脱氢酶,钾-激活 来源于面包酵母(酿酒酵母)
UFC40GV
Ultrafree®离心过滤管,样品体积2 mL
Bel-Art H37862-0202