Key Documents
AXYSCT050SSOS
Self standing screw cap tubes and attached caps with O-rings
capacity 0.5 mL, orange, sterile
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.22
材料
orange
screw top cap
無菌
sterile
特點
RCF 20,000 × g
包裝
pack of 500 tubes
case of 8 packs
製造商/商標名
Axygen Inc
Corning
容量
0.5 mL