特點
Asseptic Water Sampling
包裝
pkg of 1 unit
相容性
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
一般說明
该附件套件允许在无菌条件下对循环水进行采样。无菌取样阀用于从Milli-Q® IQ 7003/5/10/15和Milli-Q® IX 7003/5/10/15再循环回路取样。
應用
无菌取样阀用于从Milli-Q® IQ 7003/5/10/15和Milli-Q® IX 7003/5/10/15再循环回路取样。
法律資訊
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany