HP01PBVN0
1 of 10
Millipore
HP01PPVN0
Optiseal Sanitary Cartridge Housing In-line
YY1801020
Sanitary Pressure Gauge 0-7 bar (0-100 psi)
LAGR04TP1
Optiseal 0.2 µm 4′ 滤芯
LAGRM1S02
Aervent® 0.2 µm, Optiseal® 小柱
H0MMMUSAC
Millidisk Cartridge Housing In-line 40stk
CESYF0S04015
带柱塞排水孔的OPTISEAL管道过滤YF0S0415
MCGL30S03
Millidisk®小柱,带Durapore® 0.22 µm滤膜
H4VVTT000
Cartridge Housing PTFE Vent Tips
LAGB04TP6
Optiseal Durapore 筒式过滤器 0.22 µm 疏水性
HP01PPVT0
Optiseal Sanitary Cartridge Housing T-line
316L stainless steel
100
Filter Housings
Filtration
Filtration for Bioprocessing
Sterile Filtration and Bioburden Control