FT3437300WA
1 of 10
Millipore
96100014
Vantage L Seal/Bed Support Kit
96100006
Vantage L Column Tube Extension Kit
31138516WB
gasket, u section seal, id 1363mm, epdm peroxyde cured
96100050
Vantage L Tube Kit 250 mm
Sigma-Aldrich
Z556009
Aldrich® aspirator bottle with tubulation
Supelco
21003-U
Therm-O-Ring 密封圈
96100033
Vantage L Maintenance Kit
96100010
10477-U
Carbopack®吸附剂
96100041
SP2
Bioprocessing Systems
Chromatography
Chromatography Columns for Bioprocessing
Pharma & Biopharma Manufacturing