CR0003L108
1 of 10
Millipore
CR0003L102
Applikon型3L马达适配器
CR0003L200
一次性3L搅拌槽生物反应器(绿色)
BRZMBRCFLEX4
Mobius® Breez 2 mL微型生物反应器
BRZ01SYS
Mobius® Breez System
CR0003L103
Sartorius BIOSTAT A+ (1L, 2L, 5L) style 3L motor adapter
CR0003L105
Bellco CellTrol 3L with Digital OHD style 3L motor adapter
Sigma-Aldrich
P4417
Phosphate buffered saline
CLS6749
Corning®细胞计数仪
CR0003L107
New Brunswick CelliGen® Plus Style 3L motor adapter
D8537
杜氏磷酸盐缓冲盐水
EQ1