87009023
1 of 9
Millipore
87009018
Vantage S2 and A2 Column Chrom Tubes V90
87009019
Vantage S2 and A2 Routine Maintenance Kit V90
87009015
Vantage S2 and A2 Flow Cell Kit V90
MPHL02GH2
Millipak®囊式过滤器,带Durapore® 0.45 µm滤膜
87006015
Vantage S2 and A2 Flow Cell Kit V60
50652-U
Ascentis® Express 90 Å PCS-C18, 2.7 µm HPLC Column
Supelco
50536-U
Ascentis® Express C18,5μm HPLC色谱柱
Milli-Q
ZSQ240UCTK
Milli-Q® SQ 240C纯水系统套件
Sigma-Aldrich
538540
MMP-7, Proenzyme, Human, Recombinant, E. coli
Vantage
protein purification: suitable