主要文件
1.67710
Supelco
Continous level sensor for organic solvents in 10l stainless steel barrels
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122409
NACRES:
NA.03
儲存溫度
no temp limit
一般說明
包括 G2 螺纹接头、滴管、图形显示器和电缆(长度:5m)
Continuous level sensor technology with ATEX approval provides maximum safety and reliability for your daily work. The graphical display always shows the exact solvent level inside the container. With adjustable alarm, individual management, measurement and control of solvent supply and disposal now are available and automatable, e.g. by visible control alarms or automatical control of HPCL instruments.