1.50170
1 of 10
Supelco
1.50234
LiChroCART™ 55-2
1.50251
Purospher® STAR RP-18 封端(5 µm)LiChroCART™ 125-4
1.50981
LiChrospher® 60 RP-select B (5 µm) LiChroCART™ 125-4
1.50477
LiChrospher® 100 RP-18 (5 µm) Hibar® RT 125-4
1.50828
LiChrospher® 100 RP-18封尾 (5 µm) LiChroCART™ 125-4
1.50829
LiChrospher® 60 RP-select (5 µm) LiChroCART™ 125-4
1.50414
Purospher® STAR RP-18封尾 (3µm) Hibar® RT 150-3
1.51487
Pre-column holder for direct coupling of LiChroCART™ 4-4 pre-columns with Hibar® RT columns
1.51486
manu-CART™ NT卡套,用于LiChroCART™ 2,3,4,4.6 mm i.d.HPLC 柱
1.50036
Purospher® STAR RP-18 封端(5 µm) Hibar® 125-4
LiChroCART™ 125-4, HPLC cartridge
100