主要文件
Z742443
Incu-Shaker Mini dedicated platform
for 8 x 250 mL Erlenmeyer flasks
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103811
NACRES:
NB.73
特點
for 8 x 250 mL Erlenmeyer flasks
包裝
pkg of 1
製造商/商標名
Benchmark Scientific H1000-P-250
正在寻找类似产品? 访问 产品对比指南
一般說明
Dedicated platform for the Incu-Shaker Mini that features a non-slip rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays.
法律資訊
Incu-Shaker is a trademark of Benchmark Scientific, Inc.
历史批次信息供参考:
分析证书(COA)