主要文件
Z682861
Ace receiving flask,
capacity 1000 mL, joint: ST/NS 35/20, plastic-coated glass
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
plastic-coated glass
round bottom flask
single-neck
接合處
joint: ST/NS35/20
無菌
non-sterile
特點
closure type Standard Taper Joint
包裝
case of 1
製造商/商標名
Büchi equivalent, 40775/20728
容量
1000 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.
历史批次信息供参考:
分析证书(COA)