主要文件
Z678023
Finnpipette® F2 multichannel pipette
volume 1-10 μL, 12 channel
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
特點
12-channel
color-coded pink
electronic: no
包裝
pack of 1 ea
製造商/商標名
Finn 4662040
容積
1-10 μL
正在寻找类似产品? 访问 产品对比指南
一般說明
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50 μl and below
法律資訊
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
历史批次信息供参考:
分析证书(COA)