主要文件
Z677914
Finntip® Flex pipette tips
Finntip Flex 1000, non-sterile
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
最大體積:
1 mL
容積範圍:
100-1000 μL
無菌:
non-sterile
特點:
barrier: no
包裝:
pack of 1000 ea (Bag)
製造商/商標名:
Finn 94060720
材料
blue
colorless tip
無菌
non-sterile
特點
barrier: no
包裝
pack of 1000 ea (Bag)
製造商/商標名
Finn 94060720
最大體積
1 mL
容積範圍
100-1000 μL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
  • Finntip® Flex pipette tips are compatible with both single and multichannel pipettes.
  • Tips are DNA, DNase, RNase, and endotoxin-free.
  • Offers consistent seal for better precision and accurate results.
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
其他說明
Browse all pipette tips
法律資訊
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
从最新的版本中选择一种:
分析证书(COA)