主要文件
Z676470
Double-Safe PTFE coated thermometer
greener alternative
L 305 mm, temperature -20-150 °C, immersion level, total
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41112303
NACRES:
NB.79
特點
immersion level total
包裝
pack of 1 ea
製造商/商標名
HB Instruments B60400-1800
環保替代產品特色
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
溫度測量範圍
-20-150 °C
長度
305 mm
環保替代類別
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Double-Safe thermometers are PTFE coated Easy-Read thermometers with black Enviro-Safe liquid and yellow back lead free glass. Double-Safe thermometers prevent contamination of the glass and liquid in the event of breakage. Double-Safe is the ideal replacement for mercury thermometers banned (or partially banned) in many US states and most European countries.
  • PTFE coating contains liquid and glass in case of breakage
  • Non-toxic, non-hazardous, biodegradable black Enviro-Safe liquid
  • Available with statement of accuracy traceable to NIST and DKD
  • Lead-free glass and recyclable packaging
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical without mercury. Click here for more information.
法律資訊
Double-Safe is a trademark of H-B Instrument Co.
相關產品
产品编号
说明
价格
历史批次信息供参考:
分析证书(COA)