主要文件
Z657212
IKA® overhead stirrer accessories
R 2723 telescopic stand
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122800
NACRES:
NB.72
包裝
pack of 1 ea
製造商/商標名
IKA 0001412100
相关类别
一般說明
Stable with H-shape base which prevents the stand from tipping backwards. Provides optimum stability required for larger, heavier instruments and attachments. The stand has an adjustment screw which can be compensated for an uneven laboratory table surface. Additionally equipped with a pneumatic spring stand rod, which enables heavy instruments/attachments to be raised and lowered smoothly and without difficulty.
法律資訊
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
历史批次信息供参考:
分析证书(COA)
访问文档库
Global Trade Item Number
货号GTIN
Z657212-1EA4061826687352