主要文件
Z648558
Stuart Scientific SA8 vortex mixer accessories
accessory heads
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103814
NACRES:
NB.73
製造商/商標名
Bibby
存貨情形
not available in USA and Canada
正在寻找类似产品? 访问 产品对比指南
一般說明
package of
  • 1 × multiwell plate holder
  • 1 × circular foam insert for beakers and flasks up to 500 mL
  • 1 × rectangular foam insert (holds 8 x 0.2 mL, 8 x 0.5 mL and 16 x 1.5 mL microcentrifuge tubes)
  • 1 × rectangular foam insert (plain, user may cut holes to own requirements)
法律資訊
Stuart is a trademark of Bibby Sterlin Ltd.
历史批次信息供参考:
分析证书(COA)