Merck
Z513113
Sigma-Aldrich
Aldrich® non-corrosive lecture-bottle gas regulator
delivery pressure 0-100 psig, CGA Inlet size 170
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41100000
NACRES:
NB.51
delivery pressure measuring range
0-100 psig
CGA入口尺寸
170
正在寻找类似产品? 访问 产品对比指南
一般說明
  • Single-stage
  • Inlet: CGA 170
  • Outlet: 1/4 NPT Male
  • Max. inlet pressure: 3,000 psig
  • Temp. range: -40 to 140°F
  • Compact chrome-plated brass body
  • Lecture-bottle gas regulators include a PTFE sealing washer
  • Order spare PTFE sealing washers separately
其他說明
product is not intended for medical use
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)