製造商/商標名
J. Young
側臂外徑
13 mm
旋塞尺寸
10 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
Features:
- PTFE piston with unique PTFE-sheathed elastomer O-ring seals
- Grease-free glass-to-PTFE seals
- High-vacuum to 10-6 torr
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z405582-1EA | 4061826668757 |