主要文件
Z278319
Conical reaction vial
capacity 8 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121700
NACRES:
NB.43
材料
colorless tube
glass tube
screw top neck
無菌
non-sterile
特點
graduations: no
External thread
包裝
pack of 1 ea
製造商/商標名
Ace Glass D104154
容量
8 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
Thin-wall with threaded outer 14/10 joint, cap with hole, PTFE-faced silicone rubber septum, and FETFE® O-ring.
The product does NOT have the side arm opening. It is the product shown on the left of the photo.
法律資訊
FETFE is a registered trademark of Ace Glass, Inc.
历史批次信息供参考:
分析证书(COA)