主要文件
Z261300
Stainless steel 316 syringe needle, noncoring point
gauge 20, L 12 in.
登录查看公司和协议定价
About This Item
分類程式碼代碼:
42142600
NACRES:
NB.03
描述
Micro-Mate Hub with 316 Stainless Steel Tubing
包裝
pack of 1 ea
製造商/商標名
Popper 9865
長度
12 in.
測量計
20
正在寻找类似产品? 访问 产品对比指南
一般說明
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)