一般說明
Natural PP, stackable. Steam autoclavable to 250°F (121°C). Rows lettered and numbered for easy identification. Base plate has rounded wells with drain holes; end plates with hand grips.
法律資訊
Scienceware is a registered trademark of SP Scienceware, a division of SP
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z178063-1EA | 4061832262598 |