主要文件
Z174971
Round bottom flask
capacity 5 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
borosilicate glass
round bottom flask
single-neck
無菌
non-sterile
特點
closure type Screw cap
包裝
pack of 1
製造商/商標名
Ace Glass 9592-04
容量
5 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
With cap, FETFE® O-ring, and septum. Threaded <ST/>14/10 joint.
Note: This product is part of a kit. Please refer to the Z510297 web page for additional information.
法律資訊
FETFE is a registered trademark of Ace Glass, Inc.
历史批次信息供参考:
分析证书(COA)