主要文件
Z174513
Sigma-Aldrich
Aldrich® micro short-path distillation apparatus
Standard head
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41104812
NACRES:
NB.43
正在寻找类似产品? 访问 产品对比指南
一般說明
Threaded 14/20 joints. Thermometers are included.
Replacement parts:
Z174521 - Standard head
Z247774 - Jacketed head
Z174548 - Cow receiver
Z174556 - Recovery flask, 5 mL
Z174564 - Distillation flask, 10 mL
Z174572 - Gas-inlet tube
Z561487 - Thermometer, 25 mm immr. L, -10 to 250 °C
聯結
See the Threaded Ground Glass Tutorial to see how threaded joints work.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)