一般說明
用于在软木塞和橡胶塞上打孔。由带精细磨光刃口的镀锌黄铜管组成。光滑的塑料手柄刻有尺寸(3/16、1/4、5/16、3/8、7/16和1/2英寸),对应下表
请注意:每套配备一根捅条(punch)。
请注意:每套配备一根捅条(punch)。
Robert M Q Shanks et al.
PloS one, 7(5), e36398-e36398 (2012-05-23)
Serratia marcescens is a common contaminant of contact lens cases and lenses. Hemolytic factors of S. marcescens contribute to the virulence of this opportunistic bacterial pathogen. We took advantage of an observed hyper-hemolytic phenotype of crp mutants to investigate mechanisms
Bruno Pok Man Ngou et al.
Journal of experimental botany, 71(6), 2186-2197 (2020-02-13)
Plant nucleotide-binding domain, leucine-rich repeat receptor (NLR) proteins play important roles in recognition of pathogen-derived effectors. However, the mechanism by which plant NLRs activate immunity is still largely unknown. The paired Arabidopsis NLRs RRS1-R and RPS4, that confer recognition of
实验方案
This protocol covers three methods for the microscopic examination of cell samples.
This protocol covers three methods for the microscopic examination of cell samples.
Global Trade Item Number
货号 | GTIN |
---|---|
Z165220-1SET | 4061832330679 |