Z111538
1 of 10
Sigma-Aldrich
Z529672
用于数字 Wig-L-Bug® 研磨装置/混合装置的螺旋式胶囊形容器
Z111554
钢样品瓶和球形研杵,用于模拟 Wig-L-Bug® 研磨器/混合器
X6250
叶黄素
712124
反 -1-苯基-1,3-丁二烯
BAF372500010
Micro-Mill™ II grinder
424633
偶氮苯
D95654
富马酸二乙酯
205699
碳负载钯催化剂
469521
β-桧酚酮
793418
Phenyl-α-MIDA-boryl aldehyde
not available in EU
2 in.
1.0 mL
1/2 in.
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.