推荐产品
接合處
joint: ST/NS19/22
相关类别
一般說明
The Diazald kit is a set of distillation glassware designed for the safe preparation of diazomethane (~100 mmol). Glassware without sharp edges or ground-glass joints is recommended by de Boer and Backer [Org. Syn., Coll. Vol. 4, 250 (1963)]. The Diazald kit features 19/22 Clear-Seal joints; these joints do not require grease even for vacuum applications, thus avoiding a source of contamination.
The glassware in the kit should be washed with care and without the use of wire brushes which will scratch the inner surface. For further information, request Technical Information Bulletin No. AL-180 and for a review on the preparation and reactions of diazomethane, see Aldrichim. Acta, 16, 1, 1983.
Call or visit our website for pricing on replacement parts.
For technical and safety information about Diazald and Aldrich diazomethane generators see Technical Information Bulletin AL-180.
NOTE: We are unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
The glassware in the kit should be washed with care and without the use of wire brushes which will scratch the inner surface. For further information, request Technical Information Bulletin No. AL-180 and for a review on the preparation and reactions of diazomethane, see Aldrichim. Acta, 16, 1, 1983.
Call or visit our website for pricing on replacement parts.
For technical and safety information about Diazald and Aldrich diazomethane generators see Technical Information Bulletin AL-180.
NOTE: We are unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
法律資訊
Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany
试剂盒组分也可单独购买
产品编号
说明
化学品安全说明书
- Z100307Aldrich® West condenser w/ Clear-Seal joint, joint: ST/NS 19/22, pkg of 1 ea化学品安全说明书
- Z100293Aldrich® bent vacuum-distilling adapter, w/ Clear-Seal joints, joint: ST/NS 19/22化学品安全说明书
- Z100269Aldrich® Claisen adapter, with Clear-Seal joints, bottom and side joint: ST/NS 19/22, top joint: ST/NS 19/22化学品安全说明书
- Z100277Aldrich® three-way adapter with Clear-Seal joints, With Clear-Seal joints, joint: ST/NS 19/22化学品安全说明书
- Z100315Distilling column, L 200 mm, joint: ST/NS 19/22 Clear-Seal joints化学品安全说明书
- Z122513Gas-inlet tube for Aldrich® Glassware kits, pkg of 1 ea化学品安全说明书
- Z100323Aldrich® single-neck round-bottom flask with Clear-Seal joint, capacity 25 mL化学品安全说明书
- Z100331Aldrich® single-neck round-bottom flask with Clear-Seal joint, capacity 50 mL化学品安全说明书
- Z100358Aldrich® single-neck round-bottom flask with Clear-Seal joint, capacity 100 mL化学品安全说明书
- Z100366Aldrich® single-neck round-bottom flask with Clear-Seal joint, capacity 250 mL化学品安全说明书
- Z100374Aldrich® single-neck round-bottom flask with Clear-Seal joint, capacity 500 mL化学品安全说明书
- Z106283Wheaton connecting adapter, To male Clear-Seal joint, joint: ST/NS 19/22化学品安全说明书
- Z100390PTFE stopper for Macro Diazald® kit, joint: ST/NS 19/22化学品安全说明书
- Z100382Separatory funnel with PTFE stopcock and 19/22 Clear-Seal joint, capacity 125 mL化学品安全说明书
查看所有结果 (14)
更換零件
产品编号
说明
价格
其他客户在看
Joaquim Vila et al.
Applied and environmental microbiology, 75(19), 6232-6239 (2009-08-12)
The pyrene-degrading Mycobacterium sp. strain AP1 grew in nutrient-supplemented artificial seawater with a heavy fuel oil as the sole carbon source, causing the complete removal of all linear (C(12) to C(40)) and branched alkanes from the aliphatic fraction, as well
Global Trade Item Number
货号 | GTIN |
---|---|
Z100250-1KT | 4061832131146 |