主要文件
TEM-G300F1-CU
Sigma-Aldrich
Gilder Finder Grids G300F1
Copper, size 300 mesh, vial of 100
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41111761
NACRES:
MA.24
尺寸:
300 mesh
包裝:
vial of 100
包裝
vial of 100
尺寸
300 mesh
正在寻找类似产品? 访问 产品对比指南
一般說明
Gilder Finder Grids G300F1. Copper grids, 300 mesh. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 25 cells has a central alphabet letter from A to Y for identification purposes.
历史批次信息供参考:
分析证书(COA)