主要文件
SYNF100003
Synthware 47 mm filtration apparatus with fritted support
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41100000
NACRES:
NB.51
包裝
pkg of 1 ea
製造商/商標名
Synthware F100003
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has a sealed-in coarse porosity fritted disc. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
法律資訊
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)