主要文件
MT51340013
Mettler Toledo DX series GSE half-cell membrane kit
Ammonium GSE
登录查看公司和协议定价
About This Item
分類程式碼代碼:
39121436
NACRES:
NB.61
製造商/商標名
Mettler-Toledo 51340013
一般說明
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE! Kit contains one membrane plus electrolyte.
历史批次信息供参考:
分析证书(COA)