Merck
DWK92812G-1000
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask, glass
glass stopper, borosilicate glass flask, class A
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
borosilicate glass flask
glass stopper
特點
class A
包裝
case of 1 ea
製造商/商標名
Kimble® 92812G-1000
培養瓶容量
1000 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
Class A Heavy Duty Wide Mouth Volumetric Flasks with Glass Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
特點和優勢
  • Manufactured from borosilicate glass 3.3
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Heavy duty walls provide superior strength, durability and safety
  • Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Printed large volume numbers allow quick identification when selecting glassware
  • Flat interior bottoms for stirring with standard stir bars
  • Calibrate to contain
  • Graduated to Class A volumetric tolerances
法律資訊
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
历史批次信息供参考:
分析证书(COA)